Trace Metal Analysis Reagents
J.T.Baker® and Macron Fine Chemicals™ brand high purity solvents and reagents give you the performance you need - minimising the risk of contaminants that can limit accuracy while maximising instrument sensitivity and detection power in key applications.
J.T.Baker® ULTREX II Ultrapure Acids
Recommended for use in your most demanding trace element analyses; analysed for up to 65 trace elements in the low ppt range...
J.T.Baker® BAKER INSTRA-ANALYZED Acids
Recommended for ICP-OES/AES and GFAA applications, and other applications requiring ppb trace metal testing...
J.T.Baker® BAKER ANALYZED ACS Reagent Acids
Recommended for qualitative AAS applications, as well as general wet chemistry...
Macron Fine Chemicals™ brand acids
Macron Fine Chemicals™ acids generate highly reliable and consistent ppb trace metal analysis results...