Imprimer
Anti-IL7R Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: A7R34]
Fournisseur: Tonbo Biosciences
60-1271-U100 60-1271-U025
60-1271-U100EA 228 EUR
60-1271-U100 60-1271-U025
Anti-IL7R Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: A7R34]
Anticorps
The A7R34 antibody is specific for mouse CD127, a 60 to 90 kDa cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cellMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR