Imprimer
Anti-ITGAX Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: N418]
Fournisseur: Tonbo Biosciences
60-0114-U100 60-0114-U025
60-0114-U100EA 152 EUR
60-0114-U100 60-0114-U025
Anti-ITGAX Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: N418]
Anticorps
The N418 antibody reacts with mouse CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18)More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR