Imprimer

Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin))

Fournisseur: Tonbo Biosciences
50-4732-U100
50-4732-U100EA 109 EUR
50-4732-U100
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin))
Anticorps
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: MPC-11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR