Imprimer
Anti-CD11b Rat Monoclonal Antibody (In Vivo Ready™) [clone: M1/70]
Fournisseur: Tonbo Biosciences
40-0112-U100
40-0112-U100EA 108 EUR
40-0112-U100
Anti-CD11b Rat Monoclonal Antibody (In Vivo Ready™) [clone: M1/70]
Anticorps
The M1/70 antibody reacts with human and mouse CD11b, also known as integrin alpha M. This 165 to 170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles withMore Product Information
Plus About VWR