Imprimer
Click-On In-Line Super Clean® Replacement Gas Traps
Fournisseur: Restek
22464 22465
REST22464EA 228 EUR
REST22464 REST22465
Click-On In-Line Super Clean® Replacement Gas Traps
Filtres Filtres à gaz
Super-clean click-on traps measure to be 21,6×3,2 cm (8,5×1,25").
  • Triple trap - removes oxygen, moisture, and hydrocarbons