Imprimer
Human Perforin ELISPOT Kit
Fournisseur: Abcam
AB62943-96 AB62943-5X96
ABCAAB62943-96EA 632 EUR
ABCAAB62943-96 ABCAAB62943-5X96
Human Perforin ELISPOT Kit
Tests Tests immunologiques
Human Perforin ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human Perforin production in a single cell suspension.