Human IL-2 ELISA Set (without plates)
Fournisseur: Abcam
AB48471-480
AB48471-10X96
AB48471-15X96
ABCAAB48471-480EA
1140
EUR
ABCAAB48471-480
ABCAAB48471-10X96
ABCAAB48471-15X96
Human IL-2 ELISA Set (without plates)
Tests
Tests ELISA
Human IL-2 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-2 ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.