Human IL-15RA ELISA Kit
Fournisseur: Abcam
Human IL-15RA ELISA kit is a single-wash 90-min Simplestep used to quantify Human IL-15RA with a sensitivity of 5.79 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.