Imprimer
Human IRAK-1 ELISA Kit
Fournisseur: Abcam
AB219630-96 AB219630-10X96 AB219630-1X384
ABCAAB219630-96EA 969 EUR
ABCAAB219630-96 ABCAAB219630-10X96 ABCAAB219630-1X384
Human IRAK-1 ELISA Kit
Tests Tests ELISA
Human IRAK-1 ELISA kit is a single-wash 90-min Simplestep used to quantify Human IRAK-1 with a sensitivity of 22 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.