Imprimer

Anti-CCR6 Rabbit Monoclonal Antibody [clone: EPR24590-123] (Alexa Fluor® 647)

Fournisseur: Abcam
AB322226-10UL AB322226-100UL
ABCAAB322226-10ULEA 238 EUR
ABCAAB322226-10UL ABCAAB322226-100UL
Anti-CCR6 Rabbit Monoclonal Antibody [clone: EPR24590-123] (Alexa Fluor® 647)
Anticorps
Alexa Fluor® 647 Rabbit monoclonal [EPR24590-123] to CCR6.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: EPR24590-123
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR