Imprimer

Anti-Glucose Oxidase Rabbit Monoclonal Antibody [clone: EPR27335-25]

Fournisseur: Abcam
AB314545-20UL AB314545-100UL AB314545-1ML
ABCAAB314545-20ULEA 244 EUR
ABCAAB314545-20UL ABCAAB314545-100UL ABCAAB314545-1ML
Anti-Glucose Oxidase Rabbit Monoclonal Antibody [clone: EPR27335-25]
Anticorps
Rabbit monoclonal [EPR27335-25] to Glucose Oxidase.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glucose Oxidase
Clonality: Monoclonal
Clone: EPR27335-25
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR