Imprimer

Anti-RanBP2 Rabbit Recombinant Multiclonal Antibody [clone: RM2047]

Fournisseur: Abcam
AB316337-1MG AB316337-100UG
ABCAAB316337-1MGEA 9160 EUR
ABCAAB316337-1MG ABCAAB316337-100UG
Anti-RanBP2 Rabbit Recombinant Multiclonal Antibody [clone: RM2047]
Anticorps
Rabbit recombinant multiclonal [RM2047] to RanBP2 - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RanBP2
Clonality: Recombinant Multiclonal
Clone: RM2047
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR