Imprimer

Anti-V5 tag Rabbit Recombinant Multiclonal Antibody

Fournisseur: Abcam
AB314556-1MG AB314556-100UG
ABCAAB314556-1MGEA 9160 EUR
ABCAAB314556-1MG ABCAAB314556-100UG
Anti-V5 tag Rabbit Recombinant Multiclonal Antibody
Anticorps
Rabbit recombinant multiclonal [RM1083] to V5 tag - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: V5 tag
Clonality: Recombinant Multiclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR