Imprimer

Anti-EBF1 Rabbit Monoclonal Antibody [clone: EPR27344-49]

Fournisseur: Abcam
AB315450-20UL AB315450-100UL AB315450-1ML
ABCAAB315450-20ULEA 0 EUR
ABCAAB315450-20UL ABCAAB315450-100UL ABCAAB315450-1ML
Anti-EBF1 Rabbit Monoclonal Antibody [clone: EPR27344-49]
Anticorps
Rabbit monoclonal [EPR27344-49] to EBF1.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EBF1
Clonality: Monoclonal
Clone: EPR27344-49
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR