Imprimer

Anti-NAP1L1 Rabbit Recombinant Multiclonal Antibody [clone: RM1101]

Fournisseur: Abcam
AB315247-1MG AB315247-100UG
ABCAAB315247-1MGEA 9160 EUR
ABCAAB315247-1MG ABCAAB315247-100UG
Anti-NAP1L1 Rabbit Recombinant Multiclonal Antibody [clone: RM1101]
Anticorps
Rabbit recombinant multiclonal [RM1101] to NAP1L1 - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: NAP1L1
Clonality: Recombinant Multiclonal
Clone: RM1101
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR