Imprimer

Anti-C3a R Mouse Monoclonal Antibody [clone: hC3aRZ8]

Fournisseur: Abcam
AB317633-100UG AB317633-1MG
ABCAAB317633-100UGEA 1020 EUR
ABCAAB317633-100UG ABCAAB317633-1MG
Anti-C3a R Mouse Monoclonal Antibody [clone: hC3aRZ8]
Anticorps
Mouse monoclonal [hC3aRZ8] to C3a R - BSA and Azide free.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C3a R
Clonality: Monoclonal
Clone: hC3aRZ8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR