Imprimer

Anti-Uteroglobin Rabbit Monoclonal Antibody [clone: EPR27144-86] (Alexa Fluor® 647)

Fournisseur: Abcam
AB317531-10UL AB317531-100UL
ABCAAB317531-10ULEA 238 EUR
ABCAAB317531-10UL ABCAAB317531-100UL
Anti-Uteroglobin Rabbit Monoclonal Antibody [clone: EPR27144-86] (Alexa Fluor® 647)
Anticorps
Alexa Fluor® 647 Rabbit monoclonal [EPR27144-86] to Uteroglobin.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Uteroglobin
Clonality: Monoclonal
Clone: EPR27144-86
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR