Imprimer
Anti-ATP6V0D2 Rabbit Monoclonal Antibody [clone: EPR29173-521]
Fournisseur: Abcam
AB321809-20UL AB321809-100UL AB321809-1ML
ABCAAB321809-20ULEA 244 EUR
ABCAAB321809-20UL ABCAAB321809-100UL ABCAAB321809-1ML
Anti-ATP6V0D2 Rabbit Monoclonal Antibody [clone: EPR29173-521]
Anticorps
Rabbit monoclonal [EPR29173-521] to ATP6V0D2.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: ATP6V0D2
Clonality: Monoclonal
Clone: EPR29173-521
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse