Imprimer

Anti-DNA Polymerase lambda Rabbit Monoclonal Antibody [clone: EPR7519(2)]

Fournisseur: Abcam
AB172974-100 AB172974-1ML
ABCAAB172974-100EA 732 EUR
ABCAAB172974-100 ABCAAB172974-1ML
Anti-DNA Polymerase lambda Rabbit Monoclonal Antibody [clone: EPR7519(2)]
Anticorps
Rabbit monoclonal [EPR7519(2)] to DNA Polymerase lambda.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DNA Polymerase lambda
Clonality: Monoclonal
Clone: EPR7519(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR