Imprimer

Anti-SARS-CoV-2 nucleocapsid protein Human Monoclonal Antibody [clone: F12]

Fournisseur: Abcam
AB286217-100 AB286217-20UL AB286217-1ML
ABCAAB286217-100EA 744 EUR
ABCAAB286217-100 ABCAAB286217-20UL ABCAAB286217-1ML
Anti-SARS-CoV-2 nucleocapsid protein Human Monoclonal Antibody [clone: F12]
Anticorps
Human monoclonal [F12] to SARS-CoV-2 nucleocapsid protein.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 nucleocapsid protein
Clonality: Monoclonal
Clone: F12
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR