Imprimer

Anti-C1q Rat Monoclonal Antibody [clone: 7H8]

Fournisseur: Abcam
AB255973-100 AB255973-1
ABCAAB255973-100EA 1020 EUR
ABCAAB255973-100 ABCAAB255973-1
Anti-C1q Rat Monoclonal Antibody [clone: 7H8]
Anticorps
Rat monoclonal [7H8] to C1q - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C1q
Clonality: Monoclonal
Clone: 7H8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse; Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR