Imprimer

Anti-ISG15 Rabbit Monoclonal Antibody [clone: EPR24482-49]

Fournisseur: Abcam
AB285367-100 AB285367-20UL AB285367-1ML
ABCAAB285367-100EA 713 EUR
ABCAAB285367-100 ABCAAB285367-20UL ABCAAB285367-1ML
Anti-ISG15 Rabbit Monoclonal Antibody [clone: EPR24482-49]
Anticorps
Rabbit monoclonal [EPR24482-49] to ISG15.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: EPR24482-49
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR