Imprimer

Anti-GABA A Receptor alpha 5/GABRA5 Rabbit Monoclonal Antibody [clone: EPR24003-118]

Fournisseur: Abcam
AB259880-100 AB259880-20UL AB259880-1ML
ABCAAB259880-100EA 663 EUR
ABCAAB259880-100 ABCAAB259880-20UL ABCAAB259880-1ML
Anti-GABA A Receptor alpha 5/GABRA5 Rabbit Monoclonal Antibody [clone: EPR24003-118]
Anticorps
Rabbit monoclonal [EPR24003-118] to GABA A Receptor alpha 5/GABRA5.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GABA A Receptor alpha 5/GABRA5
Clonality: Monoclonal
Clone: EPR24003-118
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR