Imprimer

Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22250-300]

Fournisseur: Abcam
AB256389-1 AB256389-100
ABCAAB256389-1EA 9320 EUR
ABCAAB256389-1 ABCAAB256389-100
Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22250-300]
Anticorps
Rabbit monoclonal [EPR22250-300] to Angiotensin Converting Enzyme 1 - BSA and Azide free (Detector).

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Angiotensin Converting Enzyme 1
Clonality: Monoclonal
Clone: EPR22250-300
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR