Imprimer

Anti-ZNF217 Rabbit Monoclonal Antibody [clone: EPR6362]

Fournisseur: Abcam
AB240008-1 AB240008-100
ABCAAB240008-1EA 9160 EUR
ABCAAB240008-1 ABCAAB240008-100
Anti-ZNF217 Rabbit Monoclonal Antibody [clone: EPR6362]
Anticorps
Rabbit monoclonal [EPR6362] to ZNF217 - BSA and Azide free.

Type: Primary
Antigen: ZNF217
Clonality: Monoclonal
Clone: EPR6362
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR