Imprimer
Anti-GABA A Receptor alpha 2/GABRA2 Rabbit Monoclonal Antibody [clone: EPR26485-185]
Fournisseur: Abcam
AB307359-100 AB307359-10 AB307359-20UL AB307359-1ML
ABCAAB307359-100EA 694 EUR
ABCAAB307359-100 ABCAAB307359-10 ABCAAB307359-20UL ABCAAB307359-1ML
Anti-GABA A Receptor alpha 2/GABRA2 Rabbit Monoclonal Antibody [clone: EPR26485-185]
Anticorps
Rabbit monoclonal [EPR26485-185] to GABA A Receptor alpha 2/GABRA2.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: GABA A Receptor alpha 2/GABRA2
Clonality: Monoclonal
Clone: EPR26485-185
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat