Imprimer

Anti-GABA A Receptor gamma 2/GABRG2 Rabbit Monoclonal Antibody [clone: EPR25325-5]

Fournisseur: Abcam
AB288571-100 AB288571-1
ABCAAB288571-100EA 1020 EUR
ABCAAB288571-100 ABCAAB288571-1
Anti-GABA A Receptor gamma 2/GABRG2 Rabbit Monoclonal Antibody [clone: EPR25325-5]
Anticorps
Rabbit monoclonal [EPR25325-5] to GABA A Receptor gamma 2/GABRG2 - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GABA A Receptor gamma 2/GABRG2
Clonality: Monoclonal
Clone: EPR25325-5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR