Imprimer

Anti-CD80 Rabbit Monoclonal Antibody [clone: EPR22127-251]

Fournisseur: Abcam
AB222702-100 AB222702-20UL AB222702-1ML
ABCAAB222702-100EA 719 EUR
ABCAAB222702-100 ABCAAB222702-20UL ABCAAB222702-1ML
Anti-CD80 Rabbit Monoclonal Antibody [clone: EPR22127-251]
Anticorps
Rabbit monoclonal [EPR22127-251] to CD80.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: EPR22127-251
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR