Imprimer

Anti-OGT / O-Linked N-Acetylglucosamine Transferase Rabbit Monoclonal Antibody [clone: EPR12713]

Fournisseur: Abcam
AB177941-100 AB177941-1ML
ABCAAB177941-100EA 700 EUR
ABCAAB177941-100 ABCAAB177941-1ML
Anti-OGT / O-Linked N-Acetylglucosamine Transferase Rabbit Monoclonal Antibody [clone: EPR12713]
Anticorps
Rabbit monoclonal [EPR12713] to OGT / O-Linked N-Acetylglucosamine Transferase.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: OGT / O-Linked N-Acetylglucosamine Transferase
Clonality: Monoclonal
Clone: EPR12713
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR