Imprimer
Anti-Thiophosphate ester Rabbit Monoclonal Antibody [clone: 51-8]
Fournisseur: Abcam
AB239919-1 AB239919-100
ABCAAB239919-1EA 9160 EUR
ABCAAB239919-1 ABCAAB239919-100
Anti-Thiophosphate ester Rabbit Monoclonal Antibody [clone: 51-8]
Anticorps
Rabbit monoclonal [51-8] to Thiophosphate ester - BSA and Azide free.
Type: Primary
Antigen: Thiophosphate ester
Clonality: Monoclonal
Clone: 51-8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant