Imprimer

Anti-DNA:RNA hybrid Mouse Monoclonal Antibody [clone: S9.6]

Fournisseur: Abcam
AB234957-100 AB234957-1MG
ABCAAB234957-100EA 788 EUR
ABCAAB234957-100 ABCAAB234957-1MG
Anti-DNA:RNA hybrid Mouse Monoclonal Antibody [clone: S9.6]
Anticorps
Mouse monoclonal [S9.6] to DNA:RNA hybrid.

Type: Primary
Antigen: DNA:RNA hybrid
Clonality: Monoclonal
Clone: S9.6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR