Imprimer

Anti-CD16 Rabbit Monoclonal Antibody [clone: EPR22409-124]

Fournisseur: Abcam
AB246222-100 AB246222-1ML
ABCAAB246222-100EA 713 EUR
ABCAAB246222-100 ABCAAB246222-1ML
Anti-CD16 Rabbit Monoclonal Antibody [clone: EPR22409-124]
Anticorps
Rabbit monoclonal [EPR22409-124] to CD16.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: EPR22409-124
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR