Imprimer

Anti-ATPB Mouse Monoclonal Antibody [clone: 10/ATP]

Fournisseur: Abcam
AB289890-100 AB289890-1MG AB289890-20UG
ABCAAB289890-100EA 707 EUR
ABCAAB289890-100 ABCAAB289890-1MG ABCAAB289890-20UG
Anti-ATPB Mouse Monoclonal Antibody [clone: 10/ATP]
Anticorps
Mouse monoclonal [10/ATP] to ATPB.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ATPB
Clonality: Monoclonal
Clone: 10/ATP
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR