Imprimer

Anti-GFP Rat Monoclonal Antibody [clone: 3H9]

Fournisseur: Abcam
AB255886-1 AB255886-100
ABCAAB255886-1EA 9160 EUR
ABCAAB255886-1 ABCAAB255886-100
Anti-GFP Rat Monoclonal Antibody [clone: 3H9]
Anticorps
Rat monoclonal [3H9] to GFP - BSA and Azide free.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: 3H9
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR