Imprimer

Anti-CLEC1 Rabbit Monoclonal Antibody [clone: EPR27402-251]

Fournisseur: Abcam
AB308268-1 AB308268-100
ABCAAB308268-1EA 9200 EUR
ABCAAB308268-1 ABCAAB308268-100
Anti-CLEC1 Rabbit Monoclonal Antibody [clone: EPR27402-251]
Anticorps
Rabbit monoclonal [EPR27402-251] to CLEC1 - BSA and Azide free (Detector).

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CLEC1
Clonality: Monoclonal
Clone: EPR27402-251
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR