Imprimer

Anti-SARS-CoV-2 nucleocapsid protein Human Monoclonal Antibody [clone: ABK79-E11]

Fournisseur: Abcam
AB283894-100 AB283894-1MG AB283894-20UG
ABCAAB283894-100EA 675 EUR
ABCAAB283894-100 ABCAAB283894-1MG ABCAAB283894-20UG
Anti-SARS-CoV-2 nucleocapsid protein Human Monoclonal Antibody [clone: ABK79-E11]
Anticorps
Human monoclonal [ABK79-E11] to SARS-CoV-2 nucleocapsid protein.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 nucleocapsid protein
Clonality: Monoclonal
Clone: ABK79-E11
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR