Imprimer

Anti-Drosha Rabbit Monoclonal Antibody [clone: EPR23046-123]

Fournisseur: Abcam
AB242147-100 AB242147-1ML
ABCAAB242147-100EA 719 EUR
ABCAAB242147-100 ABCAAB242147-1ML
Anti-Drosha Rabbit Monoclonal Antibody [clone: EPR23046-123]
Anticorps
Rabbit monoclonal [EPR23046-123] to Drosha.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Drosha
Clonality: Monoclonal
Clone: EPR23046-123
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR