Imprimer

Anti-SMARCD2 Rabbit Monoclonal Antibody [clone: EPR20860-251]

Fournisseur: Abcam
AB220164-100 AB220164-1ML
ABCAAB220164-100EA 782 EUR
ABCAAB220164-100 ABCAAB220164-1ML
Anti-SMARCD2 Rabbit Monoclonal Antibody [clone: EPR20860-251]
Anticorps
Rabbit monoclonal [EPR20860-251] to SMARCD2.

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SMARCD2
Clonality: Monoclonal
Clone: EPR20860-251
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR