Imprimer
Anti-SARS-CoV-2 nucleocapsid protein Mouse Monoclonal Antibody [clone: 5B9-C6]
Fournisseur: Abcam
AB282751-100 AB282751-1
ABCAAB282751-100EA 1020 EUR
ABCAAB282751-100 ABCAAB282751-1
Anti-SARS-CoV-2 nucleocapsid protein Mouse Monoclonal Antibody [clone: 5B9-C6]
Anticorps
Mouse monoclonal [5B9-C6] to SARS-CoV-2 nucleocapsid protein - BSA and Azide free.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: SARS-CoV-2 nucleocapsid protein
Clonality: Monoclonal
Clone: 5B9-C6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Commander maintenant