Imprimer

Anti-DCTN1/p150-glued Mouse Monoclonal Antibody [clone: 1/p150Glued]

Fournisseur: Abcam
AB302966-100 AB302966-10 AB302966-20UG AB302966-1MG
ABCAAB302966-100EA 694 EUR
ABCAAB302966-100 ABCAAB302966-10 ABCAAB302966-20UG ABCAAB302966-1MG
Anti-DCTN1/p150-glued Mouse Monoclonal Antibody [clone: 1/p150Glued]
Anticorps
Mouse monoclonal [1/P150GLUED] to DCTN1/p150-glued.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DCTN1/p150-glued
Clonality: Monoclonal
Clone: 1/p150Glued
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR