Imprimer

Anti-S100 alpha 6/PRA Rabbit Monoclonal Antibody [clone: EPR13084]

Fournisseur: Abcam
AB181974-100 AB181974-1ML
ABCAAB181974-100EA 688 EUR
ABCAAB181974-100 ABCAAB181974-1ML
Anti-S100 alpha 6/PRA Rabbit Monoclonal Antibody [clone: EPR13084]
Anticorps
Rabbit monoclonal [EPR13084] to S100 alpha 6/PRA.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: S100 alpha 6/PRA
Clonality: Monoclonal
Clone: EPR13084
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR