Imprimer

Anti-Histone H4 (mono methyl R92) Rabbit Monoclonal Antibody [clone: EPR21782]

Fournisseur: Abcam
AB236214-100 AB236214-1
ABCAAB236214-100EA 1020 EUR
ABCAAB236214-100 ABCAAB236214-1
Anti-Histone H4 (mono methyl R92) Rabbit Monoclonal Antibody [clone: EPR21782]
Anticorps
Rabbit monoclonal [EPR21782] to Histone H4 (mono methyl R92) - BSA and Azide free.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H4
Clonality: Monoclonal
Clone: EPR21782
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR