Imprimer

Anti-CD16 Rabbit Monoclonal Antibody [clone: EPR22409-124]

Fournisseur: Abcam
AB285158-100 AB285158-1
ABCAAB285158-100EA 938 EUR
ABCAAB285158-100 ABCAAB285158-1
Anti-CD16 Rabbit Monoclonal Antibody [clone: EPR22409-124]
Anticorps
Rabbit monoclonal [EPR22409-124] to CD16 - BSA and Azide free (Detector).

Attention: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: EPR22409-124
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR