Imprimer

Anti-Aldehyde dehydrogenase 10 Rabbit Monoclonal Antibody [clone: EPR15425(B)]

Fournisseur: Abcam
AB184171-100 AB184171-1ML
ABCAAB184171-100EA 707 EUR
ABCAAB184171-100 ABCAAB184171-1ML
Anti-Aldehyde dehydrogenase 10 Rabbit Monoclonal Antibody [clone: EPR15425(B)]
Anticorps
Rabbit monoclonal [EPR15425(B)] to Aldehyde dehydrogenase 10.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Aldehyde dehydrogenase 10
Clonality: Monoclonal
Clone: EPR15425(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR