Imprimer
Colonnes capillaires, OPTIMA® 5
Fournisseur: MACHEREY-NAGEL
726628.50 726807.25 726807.30 726807.50 726807.60 726809.25 726809.30 726809.50 726809.60 726846.10 726854.25 726857.25 726857.50 726860.25 726860.50 726863.25 726863.50 726911.25 726911.30 726911.50 726911.60 726916.10 726916.25 726916.30 726916.50 726934.15 726934.25 726934.30 726056.10 726056.15 726056.25 726056.30 726056.50 726056.60 726099.25 726099.30 726099.50 726099.60 726313.10 726313.15 726313.25 726313.30 726313.50 726313.60 726314.15 726314.25 726314.30 726314.50 726314.60 726316.25 726316.30 726316.50 726316.60 726325.15 726325.25 726325.30 726325.50 726325.60 726523.10 726523.25 726523.30 726525.10 726525.25 726525.30 726525.50 726525.60 726541.10 726541.15 726541.25 726541.30 726623.25 726623.30 726623.50 726623.60 726628.25 726628.30 726628.60 726934.50
MANA726628.50EA 1060 EUR
MANA726628.50 MANA726807.25 MANA726807.30 MANA726807.50 MANA726807.60 MANA726809.25 MANA726809.30 MANA726809.50 MANA726809.60 MANA726846.10 MANA726854.25 MANA726857.25 MANA726857.50 MANA726860.25 MANA726860.50 MANA726863.25 MANA726863.50 MANA726911.25 MANA726911.30 MANA726911.50 MANA726911.60 MANA726916.10 MANA726916.25 MANA726916.30 MANA726916.50 MANA726934.15 MANA726934.25 MANA726934.30 MANA726056.10 MANA726056.15 MANA726056.25 MANA726056.30 MANA726056.50 MANA726056.60 MANA726099.25 MANA726099.30 MANA726099.50 MANA726099.60 MANA726313.10 MANA726313.15 MANA726313.25 MANA726313.30 MANA726313.50 MANA726313.60 MANA726314.15 MANA726314.25 MANA726314.30 MANA726314.50 MANA726314.60 MANA726316.25 MANA726316.30 MANA726316.50 MANA726316.60 MANA726325.15 MANA726325.25 MANA726325.30 MANA726325.50 MANA726325.60 MANA726523.10 MANA726523.25 MANA726523.30 MANA726525.10 MANA726525.25 MANA726525.30 MANA726525.50 MANA726525.60 MANA726541.10 MANA726541.15 MANA726541.25 MANA726541.30 MANA726623.25 MANA726623.30 MANA726623.50 MANA726623.60 MANA726628.25 MANA726628.30 MANA726628.60 MANA726934.50
Colonnes capillaires, OPTIMA® 5
Colonnes pour chromatographie
Nonpolar GC phase.
  • Standard phase with large range of applications
  • Thick film columns ≥3 μm film are expecially recommended for analytes with higher concentration
Columns with 0,10 to 0,32 mm ID and films <3 μm: max. temperature for isothermal operation 340 °C, max. temperature for short isotherms in a temperature program 360 °C; 0,53 mm ID columns with films <3 μm: max. temperatures 320 and 340 °C, resp.; thick film columns with films >3 μm: max. temperatures 300 and 320 °C, resp.
5% phenyl – 95% methylpolysiloxane phase.
USP code: G27/G36.