Imprimer

Phtalamide ≥98.0% (sur base azote total)

Fournisseur: TCI

P0283-25G P0283-500G
TCIAP0283-25GEA 26.4 EUR
TCIAP0283-25G TCIAP0283-500G
Phtalamide ≥98.0% (sur base azote total)
Phtalamide
Formule: C₈H₈N₂O₂
Numéro MDL: MFCD00025478
Numéro CAS: 88-96-0
EINECS: 201-870-6
Merck Index: 14,07369
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deb7257dc6bfbf2261d3572edd5537275405a182&vid=deb7257dc6bfbf2261d3572edd5537275405a182&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR