Imprimer

4-Chlorobenzènesulfonamide ≥98.0% (sur base azote total)

Fournisseur: TCI
Attention

C1099-25G C1099-500G
TCIAC1099-25GEA 26.4 EUR
TCIAC1099-25G TCIAC1099-500G
4-Chlorobenzènesulfonamide ≥98.0% (sur base azote total)
4-Chlorobenzènesulfonamide
Formule: C₆H₆ClNO₂S
Point d'ébullition: 316 °C (1013 hPa)
Point de fusion: 144…148 °C
Numéro MDL: MFCD00007936
Numéro CAS: 98-64-6
EINECS: 202-689-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52db605651c320aa64ec5b8aede922e7d45f1e81&vid=52db605651c320aa64ec5b8aede922e7d45f1e81&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 143.0 to 147.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR