Imprimer
Motorised cryostat, CM3050 S
Fournisseur: LEICA MICROSYSTEMS
14047033536 14047033534
LEIC14047033536EA 0 EUR
LEIC14047033536 LEIC14047033534
Motorised cryostat, CM3050 S
Microtomes et groupes de microtomes Microtomes
The CM3050 S ensures efficient specimen processing by offering fast and reliable sectioning results. An innovative heat insulation system guarantees constant stable temperatures and less power consumption.
  • Convenient section thickness selection from outside the cryochamber
  • Indication of total section thickness
  • Reproducible high-quality thin sections via step motor specimen feed
  • Programmable reverse section counter
  • Spacious cryochamber, easy to clean and disinfect
  • Convenient access to the cooling system from outside the cryostat housing
  • Encapsulated
More Product Information