Imprimer
Colonnes capillaires, Elite-CLPesticides pour autosystem
Fournisseur: PerkinElmer
PERCN9316662EA 1170 EUR
PERCN9316662 PERCN9316661 PERCN9316664 PERCN9316663 PERCN9316666 PERCN9316665
Colonnes capillaires, Elite-CLPesticides pour autosystem
Colonnes pour chromatographie
Elite-CL Pesticides capillary columns are specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide.